盐酸 1 N, BAKER ANALYZED® Volumetric, J.T.Baker®

供应商: Avantor
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

M.O.C. Category NI

BAKR5620-02EA 841 CNY
BAKR5620-02 BAKR5620-07 BAKR5620-03
盐酸 1 N, BAKER ANALYZED® Volumetric, J.T.Baker®
盐酸
分子式: HCl
分子量: 36.46 g/mol
存储条件: Ambient
MDL编号: MFCD00011324
CAS号: 7647-01-0
EINECS: 231-595-7
UN号: 1789
ADR: 8 II

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (Clear, colorless liquid) Passes Test
Normality 0.995 - 1.005
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR