氯化锰(II) 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

供应商: Avantor
Warning

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

BAKR2540-07EA 0 CNY
BAKR2540-07 BAKR2540-01 BAKR2540-R BAKR2540-05
氯化锰(II) 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
氯化锰(II)
分子式: MnCl₂·4H₂O
分子量: 197.9 g/mol
沸点: 1190 °C (1013 hPa)
溶点: 58…59 °C
密度: 2.01 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00149792
CAS号: 13446-34-9
EINECS: 231-869-6
UN号: 3077
ADR: 9 III
Merck Index: 13,05751

现在订购

产品技术规格

Meets ACS Reagent Chemical Requirements 98.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
ACS - Assay (MnCl₂·4H₂O) (by EDTA titrn) 98.0 - 101.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 3.5 - 6.0
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
ACS - Zinc (Zn) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR