Dextrose kit, anhydrous USP

供应商: Avantor
Warning

Synonyms: 右旋糖

BAKR1919-X2EA 0 CNY
BAKR1919-X2
Dextrose kit, anhydrous USP
D-(+)-葡萄糖
分子式: C₆H₁₂O₆
分子量: 180.16 g/mol
溶点: 146 °C
密度: 1.54 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00148912
CAS号: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459

现在订购

产品技术规格

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Kit Components Meet Requirements Passes Test
This kit contains one each of Dextrose (Glucose) 1919-QM and 1919-SA.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR