Robotic tips, Axygen®, for PerkinElmer® Janus (Roborack)
供应商: Corning
RRP-20-C-R RRP-20-CBK-R RRP-200-C-R RRP-200-CBK-R RRP-50-C-R RRP-50-CBK-R RRF-25-C-R-S RRP-20-CBK-R-S RRP-200-C-R-S RRP-50-C-R-S RRP-50-CBK-R-S RRF-175-C-R-S RRF-25-CBK-R-S
AXYGRRP-20-C-REA 0 CNY
AXYGRRP-20-C-R AXYGRRP-20-CBK-R AXYGRRP-200-C-R AXYGRRP-200-CBK-R AXYGRRP-50-C-R AXYGRRP-50-CBK-R AXYGRRF-25-C-R-S AXYGRRP-20-CBK-R-S AXYGRRP-200-C-R-S AXYGRRP-50-C-R-S AXYGRRP-50-CBK-R-S AXYGRRF-175-C-R-S AXYGRRF-25-CBK-R-S
Robotic tips, Axygen®, for PerkinElmer® Janus (Roborack)
吸头
Transparent or black PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid-level sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase, DNase and endotoxin-free
Rack insert colour: Blue (20 µl)/green (50 µl)/red (200 µl).
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 I-RECs, ISO 14001 - Reynosa