Mini tube systems (MTS) caps, Axygen®
供应商: Corning
MTS-12CP-C MTS-12CP-C-S MTS-8CP-C MTS-8CP-C-S
AXYGMTS-12CP-CEA 0 CNY
AXYGMTS-12CP-C AXYGMTS-12CP-C-S AXYGMTS-8CP-C AXYGMTS-8CP-C-S
Mini tube systems (MTS) caps, Axygen®
外壳 检测/样品管盖
Axygen® mini tube systems are available in 8-cap or 12-cap strip format.
  • Molded-in condensation rings on the lid minimise cross contamination