Sealing mats for assay and storage microplates, Axygen®
供应商: Corning
AM-2ML-RD AM-2ML-RD-S AM-2ML-SQ AM-384-DW-SQ AM-500UL-RD AM-750UL-RD
AXYGAM-2ML-RDEA 0 CNY
AXYGAM-2ML-RD AXYGAM-2ML-RD-S AXYGAM-2ML-SQ AXYGAM-384-DW-SQ AXYGAM-500UL-RD AXYGAM-750UL-RD
Sealing mats for assay and storage microplates, Axygen®
密封件 微孔板封板膜和铝箔
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
  • Minimize evaporation
  • Made of research-grade silicone
  • Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.
现在订购
规格
Environmentally Preferable
环境认证