3-Hydroxy-2-pyrrolidinone 95%

供应商: Thermo Fisher Scientific
Warning

H57961.03
ALFAH57961.03EA 0 CNY
ALFAH57961.03
3-Hydroxy-2-pyrrolidinone 95%
3-Hydroxy-2-pyrrolidinone

Purity: 95% CAS No.: 15116-68-4 Molecular Formula: C4H7NO2 Formula Weight: 101.11 Storage Temperature: 2°C to 8°C Melting Point: 102-107° MDL No.: MFCD09751256

分子式: C₄H₇NO₂
分子量: 101.11 g/mol
溶点: 102…107 °C
MDL编号: MFCD09751256
CAS号: 15166-68-4

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR