三羟甲基氨基甲烷 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Tris

A18494.36
ALFAA18494.36EA 0 CNY
ALFAA18494.36
三羟甲基氨基甲烷 99%
三羟甲基氨基甲烷

Purity: 99% CAS No.: 77-86-1 Molecular Formula: C4H11NO3 Formula Weight: 121.14 Melting Point: 168-172° Boiling Point: 219-220°/10mm Merck Reference: 14,9772 MDL No.: MFCD00004679

分子式: NH₂C(CH₂OH)₃
分子量: 121.14 g/mol
沸点: 219…220 °C (10 mmHg)
溶点: 168…172 °C
密度: 1.353
存储条件: Ambient
MDL编号: MFCD00004679
CAS号: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR