甲酰甲撑基三苯基磷, (max. 3% H₂O) 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: (甲酰亚甲基)三苯基正膦

A11709.14
ALFAA11709.14EA 0 CNY
ALFAA11709.14
甲酰甲撑基三苯基磷, (max. 3% H₂O) 97%
甲酰甲撑基三苯基磷

Purity: 97% CAS No.: 2136-75-6 Molecular Formula: (C6H5)3P=CHCHO Formula Weight: 304.33 Storage Temperature: 2°C to 8°C Melting Point: 183-188° Beilstein Reference: 523797 MDL No.: MFCD00006994

分子式: C₂₀H₁₇OP
分子量: 304.33 g/mol
溶点: 183…188 °C
存储条件: Refrigerator
MDL编号: MFCD00006994
CAS号: 2136-75-6
EINECS: 218-375-6

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR