2,2,4-三甲基戊烷 99+% ACS for spectroscopy

供应商: Thermo Fisher Scientific
Danger

Synonyms: 异辛烷

421985000
ACRO421985000EA 0 CNY
ACRO421985000
2,2,4-三甲基戊烷 99+% ACS for spectroscopy
2,2,4-三甲基戊烷
分子式: (CH₃)₂CHCH₂C(CH₃)₃
分子量: 114.23 g/mol
沸点: 99.2
溶点: –107 °C
密度: 0.69 g/cm³
闪点: –12 °C
存储条件: Ambient
MDL编号: MFCD00008943
CAS号: 540-84-1
EINECS: 208-759-1
UN号: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear liquid
GC ≥99.0 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR