1,4-二氧六环 99+% Stabilised ACS, meetSpec_Reagent Ph.Eur.

供应商: Thermo Fisher Scientific
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

408825000
ACRO408825000EA 0 CNY
ACRO408825000
1,4-二氧六环 99+% Stabilised ACS, meetSpec_Reagent Ph.Eur.
1,4-二氧六环
分子式: C₄H₈O₂
分子量: 88.11 g/mol
沸点: 101.1
溶点: 12 °C
密度: 1.034 g/cm³
闪点: 12
存储条件: Ambient
MDL编号: MFCD00006571
CAS号: 123-91-1
UN号: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
Density (20°C) 1.032 to 1.036 g/ml
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 5 to 30 ppm BHT
Residue after evaporation ≤0.005 %
Carbonyl (as HCHO) ≤0.01 %
Titrable acid ≤0.0016 meq/g
Peroxides ≤0.005 % (H2O2)
Freezing point ≥11.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR