N,N-二甲基甲酰胺 99.8% Molecular_Biology DNAse_RNAse_PROTEase_free

供应商: Thermo Fisher Scientific
Danger

Synonyms: DMF

327175000
ACRO327175000EA 0 CNY
ACRO327175000
N,N-二甲基甲酰胺 99.8% Molecular_Biology DNAse_RNAse_PROTEase_free
N,N-二甲基甲酰胺
分子式: HCON(CH₃)₂
分子量: 73.09 g/mol
沸点: 153 °C (760 mmHg)
溶点: –61 °C
密度: 0.945 g/cm³
闪点: 58 °C
存储条件: Ambient
MDL编号: MFCD00003284
CAS号: 68-12-2
EINECS: 200-679-5
UN号: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.8 %
UV at 280 nm A: ≤0.1
Color scale ≤5 APHA
Residue after evaporation ≤0.001 %
Dnase,Rnase,Protease activity none detected
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Free amine ≤0.001 % (Kaiser)
Acidity (CH3COOH) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR