杆菌肽, potency min. 60 Units/mg

供应商: Thermo Fisher Scientific

Synonyms: 杆菌肽锌

226110050
ACRO226110050EA 0 CNY
ACRO226110050
杆菌肽, potency min. 60 Units/mg
杆菌肽
分子式: C₆₆H₁₀₁N₁₇O₁₆SZn
溶点: 250 °C (dec.)
存储条件: Refrigerator
MDL编号: MFCD28168035
CAS号: 1405-89-6

现在订购

产品技术规格

Appearance White to yellow to beige to light grey Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 %
Potency ≥60 U/mg (On dried substance)
pH 6.0 to 7.5 (10 % solution)
Zinc (Zn) 4.0 to 6.0 % (on dried substance)

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR