碱式碳酸锌 58% (53% Zn min.), powder, extra_pure

供应商: Thermo Fisher Scientific
Warning

Synonyms: 碳酸锌氢氧化物

222650010
ACRO222650010EA 0 CNY
ACRO222650010
碱式碳酸锌 58% (53% Zn min.), powder, extra_pure
碱式碳酸锌
分子式: C₂H₆O₁₂Zn₅
分子量: 549.01 g/mol
溶点: > 140 °C
密度: 4.39 g/cm³
MDL编号: MFCD00011519
CAS号: 5263-02-5
EINECS: 226-076-7
UN号: 3077

现在订购

产品技术规格

Appearance White Crystalline powder
Titration Complexometric ≥53 % (Zn)
Chloride (Cl) ≤0.3 %
Sulfate (SO4) ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR