N,N-二甲基苄胺 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: DMBA, N-苄基二甲胺, BDMA

159751000 159755000
ACRO159751000EA 0 CNY
ACRO159751000 ACRO159755000
N,N-二甲基苄胺 99%
N,N-二甲基苄胺
分子式: C₆H₅CH₂N(CH₃)₂
分子量: 135.21 g/mol
沸点: 183…184 °C (760 mmHg)
溶点: –75 °C
密度: 0.9 g/cm³
闪点: 54 °C
存储条件: Ambient
MDL编号: MFCD00008329
CAS号: 103-83-3
EINECS: 203-149-1
UN号: 2619
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=7044105a7e7fbce7e00f1d15d7a4e017a1b9d418&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5020 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %
Filt.over 0.2 micron PFTE Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR