1,5-二氮杂双环[4.3.0]壬-5-烯 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: DBN

159610250 159611000
ACRO159610250EA 0 CNY
ACRO159610250 ACRO159611000
1,5-二氮杂双环[4.3.0]壬-5-烯 98%
1,5-二氮杂双环[4.3.0]壬-5-烯
分子式: C₇H₁₂N₂
分子量: 124.19 g/mol
沸点: 95…98 °C (7.5 mmHg)
密度: 1 g/cm³
闪点: 94
存储条件: Ambient
MDL编号: MFCD00005554
CAS号: 3001-72-7
EINECS: 221-087-3
UN号: 3267
ADR: 8 II

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5210 (20°C, 589 nm)
Water ≤1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR