1,5-二苯基卡巴肼 98% ACS

供应商: Thermo Fisher Scientific
Warning

Synonyms: 1,5-二苯羰酰肼, 对称二苯氨基脲

158840250 158841000
ACRO158840250EA 0 CNY
ACRO158840250 ACRO158841000
1,5-二苯基卡巴肼 98% ACS
1,5-二苯基卡巴肼
分子式: C₁₃H₁₄N₄O
分子量: 242.28 g/mol
沸点: 454
溶点: 173…176 °C
存储条件: Ambient
MDL编号: MFCD00003013
CAS号: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance light grey to light purple to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 173 to 176 °C
Sulfated ash ≤0.05 %
Solubility (in aqueous acetone) Passes test
HPLC ≥97.5 %
Sensitivity (to chromate) Passes test

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR