苯己酮 98%

供应商: Thermo Fisher Scientific

Synonyms: 戊基苯基酮

120720250
ACRO120720250EA 0 CNY
ACRO120720250
苯己酮 98%
苯己酮
分子式: C₁₂H₁₆O
分子量: 176.26 g/mol
沸点: 265.1 °C (760 mmHg)
溶点: 25…26 °C
密度: 0.95 g/cm³
闪点: >112 °C
存储条件: Ambient
MDL编号: MFCD00009512
CAS号: 942-92-7
EINECS: 213-394-6

现在订购

产品技术规格

Appearance Clear light yellow Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR