苯甲酰肼 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 苯甲酸肼, 苯酰肼

105771000
ACRO105771000EA 0 CNY
ACRO105771000
苯甲酰肼 98%
苯甲酰肼
分子式: C₇H₈N₂O
分子量: 136.15 g/mol
溶点: 113…117 °C
存储条件: Ambient
MDL编号: MFCD00007596
CAS号: 613-94-5
EINECS: 210-363-9

现在订购

产品技术规格

Appearance White to beige Powder or crystals or needles
Infrared spectrum Conforms
Melting point 113 to 117 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR