Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 5 µm
供应商: Avantor
ACETMDKA-5-2546UEA 18143 CNY
ACETMDKA-5-2546U ACETMDKA-5-2503U ACETMDKA-5-2502U ACETMDKA-5-1546U ACETMDKA-5-1503U ACETMDKA-5-1502U ACETMDKA-5-1046U ACETMDKA-5-1003U ACETMDKA-5-1002U ACETMDKA-5-0546U ACETMDKA-5-0503U ACETMDKA-5-0502U
Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 5 µm
色谱柱
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 µm and 5 µm ) and complementary phases
  • Analytical IDs in
More Product Information