Avantor® ACE®, UHPLC Column Connectors
供应商: Avantor
ACETEXL-TWEA 9144 CNY
ACETEXL-TW ACETEXL-CCSK ACETEXL-CC10 ACETEXL-CC
Avantor® ACE®, UHPLC Column Connectors
接头
Stainless steel/PEEK UHPLC column inlet / outlet connectors. These column connectors are designed to connect UHPLC columns to UHPLC systems and make other similar connections. They are suitable for use with ¹/₁₆" ext.Ø tubing and virtually all manufacturers' UHPLC systems and all brands of UHPLC columns.
  • Pressure rated to 1700 bar (>25000 psi)
  • Usable at elevated temperatures (up to 100 °C)
  • Reusable, non-permanent swaging design
  • 10 make and break cycles
For optimum connector lifetime and to avoid accidental permanent swaging of the connector to theMore Product Information