Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 1,7 µm
供应商: Avantor
ACETEXL-1712-7502UEA 7204 CNY
ACETEXL-1712-7502U ACETEXL-1712-1003U ACETEXL-1712-1002U ACETEXL-1712-0503U ACETEXL-1712-0502U ACETEXL-1712-0302U ACETEXL-1712-0202U ACETACE-1P12-GD2U
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 1,7 µm
色谱柱
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information