Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 5 µm
供应商: Avantor
ACETEXL-129-1002UEA 6270 CNY
ACETEXL-129-1002U ACETEXL-129-1003U ACETEXL-129-1046U ACETEXL-129-1502U ACETEXL-129-1503U ACETEXL-129-1546U ACETEXL-129-2502U ACETEXL-129-2503U ACETEXL-129-2546U ACETEXL-129-0303U ACETEXL-129-0502U ACETEXL-129-0503U ACETEXL-129-0546U ACETEXL-129-7546U
Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 5 µm
色谱柱
The Avantor® ACE® Excel® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 10
More Product Information