Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 2 µm
供应商: Avantor
ACETEXL-109-1002UEA 6937 CNY
ACETEXL-109-1002U ACETEXL-109-1003U ACETEXL-109-1046U ACETEXL-109-1202U ACETEXL-109-1203U ACETEXL-109-1246U ACETEXL-109-1502U ACETEXL-109-1503U ACETEXL-109-1546U ACETEXL-109-0302U ACETEXL-109-0303U ACETEXL-109-3502U ACETEXL-109-0502U ACETEXL-109-0503U ACETEXL-109-0546U ACETEXL-109-7502U ACETEXL-109-7503U ACETEXL-109-7546U ACETEXL1091002UMVK ACETEXL1091502UMVK ACETACE-1P9-GD4U
Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 2 µm
色谱柱
The Avantor® ACE® Excel® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µ
More Product Information