Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, 2 µm
供应商: Avantor
ACETEXL-1012-0502UEA 6671 CNY
ACETEXL-1012-0502U ACETEXL-1012-0503U ACETEXL-1012-0546U ACETEXL-1012-7502U ACETEXL-1012-7503U ACETEXL-1012-7546U ACETEXL-1012-1502U ACETEXL-1012-1503U ACETEXL-1012-1546U ACETEXL-1012-1002U ACETEXL-1012-1003U ACETEXL-1012-1046U ACETEXL-1012-1202U ACETEXL-1012-1203U ACETACE-1P12-GD4U
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, 2 µm
色谱柱
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information