Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 2 µm
供应商: Avantor
ACETEXL-1010-1002UEA 6937 CNY
ACETEXL-1010-1002U ACETEXL-1010-1003U ACETEXL-1010-1046U ACETEXL-1010-1202U ACETEXL-1010-1203U ACETEXL-1010-1502U ACETEXL-1010-1503U ACETEXL-1010-1546U ACETEXL-1010-0202U ACETEXL-1010-0302U ACETEXL-1010-0502U ACETEXL-1010-0503U ACETEXL-1010-0546U ACETEXL-1010-7502U ACETEXL-1010-7503U ACETEXL-1010-7546U ACETACE-1P10-GD4U
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 2 µm
色谱柱
Avantor® ACE® Excel® C18-PFP phase utilizes a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Also useful for analytes with π-bonding or
More Product Information