VWR® ACE® UltraCore Super C18 HPLC/UHPLC Columns, 2,5 µm
供应商: Avantor
ACETCORE-25A-15005EA 8804 CNY
ACETCORE-25A-15005 ACETCORE-25A-10005 ACETCORE-25A-0301 ACETCORE-25A-03005 ACETCORE-25A-7546U ACETCORE-25A-7503U ACETCORE-25A-7502U ACETCORE-25A-3503U ACETCORE-25A-1546U ACETCORE-25A-1503U ACETCORE-25A-1502U ACETCORE-25A-1246U ACETCORE-25A-1203U ACETCORE-25A-1202U ACETCORE-25A-1046U ACETCORE-25A-1003U ACETCORE-25A-1002U ACETCORE-25A-0546U ACETCORE-25A-0503U ACETCORE-25A-0502U ACETCORE-25A-0346U ACETCORE-25A-0303U ACETCORE-25A-0302U ACETCORE-25A-0246U ACETCORE-25A-0203U ACETCORE-25A-0202U ACETCORE-A-GD2U ACETCORE-A-GD4U
VWR® ACE® UltraCore Super C18 HPLC/UHPLC Columns, 2,5 µm
色谱柱
Avantor® ACE® UltraCore SuperC18 solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. These stainless steel columns combine high efficiency separations with low column back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and effectively eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability.
  • Ideal for demanding MS applications
More Product Information