Avantor® ACE® UltraCore SuperC18, HPLC/UHPLC Columns, 5 µm
供应商: Avantor
ACETCORE-5A-7502UEA 7471 CNY
ACETCORE-5A-7502U ACETCORE-5A-2546U ACETCORE-5A-2503U ACETCORE-5A-2502U ACETCORE-5A-1546U ACETCORE-5A-1503U ACETCORE-5A-1502U ACETCORE-5A-1246U ACETCORE-5A-1046U ACETCORE-5A-1003U ACETCORE-5A-1002U ACETCORE-5A-0546U ACETCORE-5A-0503U ACETCORE-5A-0502U ACETCORE-5A-0346U ACETCORE-5A-0302U ACETCORE-5A-0202U
Avantor® ACE® UltraCore SuperC18, HPLC/UHPLC Columns, 5 µm
色谱柱
Avantor® ACE® UltraCore SuperC18 solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. These stainless steel columns combine high efficiency separations with low column back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and effectively eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability.
  • Ideal for demanding MS applications
More Product Information