2-溴-1,3-二甲苯 ≥98.0% (by GC)
供应商: TCI
Warning
Synonyms: 2-溴-间二甲苯
B0669-25G B0669-250G
TCIAB0669-25GEA 0 CNY
TCIAB0669-25G TCIAB0669-250G
2-溴-1,3-二甲苯 ≥98.0% (by GC)
2-溴-1,3-二甲苯
分子式: C₈H₉Br
沸点: 206°C
溶点: –10 °C
密度: 1,38 g/cm³ (20 °C)
闪点: 74°C
存储条件: Ambient
MDL编号: MFCD00000075
CAS号: 576-22-7
EINECS: 209-397-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1e46eabc8bee43ccd2b6f6ddd645c01efc52cdb&vid=05ba0f96106cbb2164a8810156abf5895a06ba80&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %