o,α,α,α-Tetrafluoroanisole ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
α,α,α,2-Tetrafluoroanisole, 2-(Trifluoromethoxy)fluorobenzene, 1-Fluoro-2-(trifluoromethoxy)benzene
T2150-25G
T2150-5G
TCIAT2150-25GEA
0
CNY
TCIAT2150-25G
TCIAT2150-5G
o,α,α,α-Tetrafluoroanisole ≥98.0% (by GC)
o,α,α,α-Tetrafluoroanisole
分子式:
C₇H₄F₄O 沸点: 105°C |
MDL编号:
MFCD00236322 CAS号: 2106-18-5 UN号: 1993 ADR: 3,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...