2,5-二甲氧基甲苯 ≥98.0% (by GC)
供应商: TCI
Warning
D2552-25G D2552-5G
TCIAD2552-25GEA 0 CNY
TCIAD2552-25G TCIAD2552-5G
2,5-二甲氧基甲苯 ≥98.0% (by GC)
2,5-二甲氧基甲苯
分子式: C₉H₁₂O₂
沸点: 218°C
溶点: 15 °C
密度: 1,04 g/cm³ (20 °C)
闪点: 102°C
MDL编号: MFCD00041901
CAS号: 24599-58-4
EINECS: 246-343-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5063ebd50637e5057fc84169e41b3b9798c8151a&vid=5063ebd50637e5057fc84169e41b3b9798c8151a&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %