1,4-Dipropylbenzene ≥97.0% (by GC)

供应商: TCI

D2780-25ML D2780-5ML
TCIAD2780-25MLEA 0 CNY
TCIAD2780-25ML TCIAD2780-5ML
1,4-Dipropylbenzene ≥97.0% (by GC)
1,4-Dipropylbenzene
分子式: C₁₂H₁₈
沸点: 221°C
MDL编号: MFCD03844789
CAS号: 4815-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d640a14dc940de5eb732e77d10125873f4f4c47&vid=6d640a14dc940de5eb732e77d10125873f4f4c47&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR